4-Methyl-3-(trifluoromethyl)benzonitrile 98%
Supplier: Ambeed
|
Danger
|
A265484-1G
A265484-25G
A265484-5G
A265484-100G
A265484-250MG
103812-732EA
6
USD
103812-732
103852-626
77092-316
77092-318
77092-320
77289-328
77289-330
4-Methyl-3-(trifluoromethyl)benzonitrile 98%
4-Methyl-3-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₉H₆F₃N Boiling Pt: 220 °C (760 mm Hg) Melting Pt: 60…62 °C Flash Pt: 86.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631597 CAS Number: 261952-06-1 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...