5-Amino-2-methylphenylboronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-methylphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-(5-amino-2-methylphenyl)-1,3,2-dioxaborolane, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenamine
A210996-100MG
A210996-250MG
A210996-1G
A210996-25G
A210996-5G
A210996-10G
103818-062EA
13
USD
103818-062
103848-754
76770-508
76770-510
77087-148
77087-150
77087-152
77877-662
5-Amino-2-methylphenylboronic acid pinacol ester 95%
5-Amino-2-methylphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₂₀BNO₂ Melting Pt: 82…86 °C Flash Pt: 175.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05663859 CAS Number: 882670-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...