4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid 97%

Supplier: Ambeed
Warning

A114039-250MG A114039-25G A114039-1G A114039-5G
103798-334EA 6 USD
103798-334 103824-176 77025-950 77025-952 77310-994 77310-996
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid 97%
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₄O₂
MW: 306.3187 g/mol
Boiling Pt: 587.9 °C (760 mm Hg)
Flash Pt: 309.367 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11521324
CAS Number: 641569-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR