4-Methyl-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A145179-100MG A145179-5G
103803-500EA 58 USD
103803-500 103841-582 77285-906 77316-860
4-Methyl-2-nitrobenzaldehyde 98%
4-Methyl-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₃
Boiling Pt: 309.4 °C (760 mm Hg)
Flash Pt: 154.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669895
CAS Number: 20357-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR