4-Methyl-1-naphthoic acid 97%

Supplier: Ambeed
Warning

A209848-25G A209848-5G A209848-10G A209848-100G
103848-648EA 25 USD
103848-648 77087-006 77087-008 77164-904 77892-314
4-Methyl-1-naphthoic acid 97%
4-Methyl-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
Boiling Pt: 387.4 °C (760 mm Hg)
Melting Pt: 179…181 °C
Flash Pt: 174 °C
Storage Temperature: Ambient
MDL Number: MFCD00671557
CAS Number: 4488-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR