4-Methyl-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Ambeed
|
Warning
|
A197680-100MG
A197680-250MG
103816-956EA
93
USD
103816-956
103847-578
77287-604
77287-606
4-Methyl-1H-pyrrolo[2,3-b]pyridine 95%
4-Methyl-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₈H₈N₂ Melting Pt: 125…126 °C Flash Pt: 128.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00956196 CAS Number: 824-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...