4-Methyl-1H-pyrazol-3-amine 95%

Supplier: Ambeed
Warning

A252072-1G A252072-5G A252072-100MG A252072-250MG
103812-022EA 71 USD
103812-022 103851-864 77091-264 77091-266 77289-064 77289-066
4-Methyl-1H-pyrazol-3-amine 95%
4-Methyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₄H₇N₃
Boiling Pt: 314.461 °C (760 mm Hg)
Flash Pt: 169.751 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08272068
CAS Number: 64781-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR