4-Methylindole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-1H-indole

A239855-1G A239855-25G A239855-5G A239855-100G A239855-10G
103811-314EA 5 USD
103811-314 103851-112 77090-258 77090-260 77090-262 77275-992 77275-994
4-Methylindole 97%
4-Methylindole

Precaución: for Research Use Only

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 267.8 °C (760 mm Hg)
Melting Pt: 5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005668
CAS Number: 16096-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR