4-Methylimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-1H-imidazole

A508676-1000G A508676-25G A508676-100G A508676-500G
103810-410EA 70 USD
103810-410 76843-444 76843-446 77116-604 77116-606
4-Methylimidazole 98%
4-Methylimidazole

Precaución: for Research Use Only

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 263 °C (760 mm Hg)
Melting Pt: 44…47 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00005201
CAS Number: 822-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR