4-Methylbiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-1,1'-biphenyl, 4-Phenyltoluene

A172556-100G A172556-10G A172556-25G A172556-5G A172556-500G
103845-014EA 6 USD
103845-014 76720-552 76720-554 77082-208 77082-210 77667-408
4-Methylbiphenyl 98%
4-Methylbiphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 267…268 °C (760 mm Hg)
Melting Pt: 44…48 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00008544
CAS Number: 644-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR