4-Methoxypyrimidine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A147432-250MG A147432-1G A147432-5G
103803-776EA 106 USD
103803-776 103841-878 77010-604 77317-246 77317-248
4-Methoxypyrimidine-5-carboxylic acid 95%
4-Methoxypyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20644991
CAS Number: 72411-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR