4-Methoxypiperidine 97%

Supplier: Ambeed
Warning

A106944-100G A106944-10G A106944-25G A106944-1G A106944-5G
103797-098EA 12 USD
103797-098 103822-856 76869-482 76869-484 76869-486 77309-720 77309-722
4-Methoxypiperidine 97%
4-Methoxypiperidine

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MW: 115.1700 g/mol
Boiling Pt: 149.9 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04115010
CAS Number: 4045-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR