4-Methoxypyridine-2-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-methoxypicolinonitrile
A271426-10G
A271426-250MG
A271426-1G
A271426-25G
A271426-5G
103813-046EA
17
USD
103813-046
103852-958
77289-452
77289-454
77305-072
77305-074
77305-076
4-Methoxypyridine-2-carbonitrile 97%
4-Methoxypyridine-2-carbonitrile
Precaución: for Research Use Only
Formula:
C₇H₆N₂O MW: 134.1400 g/mol Boiling Pt: 257.9 °C (760 mm Hg) Melting Pt: 118…121 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08062857 CAS Number: 36057-44-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...