2-Formyl-4-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxypicolinaldehyde, 4-Methoxypyridine-2-carboxaldehyde

A140010-100MG A140010-250MG A140010-1G A140010-25G A140010-5G
103802-812EA 30 USD
103802-812 77285-698 77285-700 77299-110 77315-920 77315-922
2-Formyl-4-methoxypyridine 98%
2-Formyl-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.1400 g/mol
Boiling Pt: 238.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08234865
CAS Number: 16744-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR