4-Methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid

A376270-100G A376270-10G A376270-500G A376270-25G A376270-5G
103837-604EA 9 USD
103837-604 76773-512 76773-514 76773-516 76840-908 77306-874
4-Methoxyphenylboronic acid 98%
4-Methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Boiling Pt: 306.8 °C (760 mm Hg)
Melting Pt: 204…206 °C
Flash Pt: 139.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR