(4-Methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxybenzyl cyanide, 4-Methoxyphenylacetonitrile

A194177-10G A194177-25G
103847-240EA 5 USD
103847-240 77029-918 77302-396
(4-Methoxyphenyl)acetonitrile 98%
(4-Methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 286…287 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001919
CAS Number: 104-47-2
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR