3,4-Diaminoanisole 97%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxy-o-phenylenediamine

A191656-1G A191656-5G A191656-100G A191656-250MG A191656-25G
103816-390EA 22 USD
103816-390 103846-994 77084-846 77084-848 77287-492 77287-494 77287-496 77287-498
3,4-Diaminoanisole 97%
3,4-Diaminoanisole

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
Boiling Pt: 302.4 °C (760 mm Hg)
Melting Pt: 46…48 °C
Flash Pt: 161.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047837
CAS Number: 102-51-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR