4-Methoxy-N-methylbenzylamine 98%

Supplier: Ambeed
Danger

A326303-1G A326303-25G A326303-5G A326303-100G A326303-10G
103825-426EA 6 USD
103825-426 103836-374 76839-324 76839-326 76839-328 77167-352 77167-354
4-Methoxy-N-methylbenzylamine 98%
4-Methoxy-N-methylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 222 °C (760 mm Hg)
Flash Pt: 85.1 °C
Storage Temperature: Ambient
MDL Number: MFCD04115407
CAS Number: 702-24-9
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR