4-Methoxy-N-methylaniline 95%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-p-anisidine

A154384-100G A154384-10G A154384-500G A154384-1G A154384-25G A154384-5G
103804-634EA 5 USD
103804-634 103842-822 76719-870 76719-872 76719-874 77318-536 77318-538 77318-540
4-Methoxy-N-methylaniline 95%
4-Methoxy-N-methylaniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 252.4 °C (760 mm Hg)
Melting Pt: 33…36 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008399
CAS Number: 5961-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR