4-Methoxynicotinic acid 95%

Supplier: Ambeed
Warning

A126188-100MG A126188-250MG A126188-1G A126188-5G
103800-770EA 38 USD
103800-770 77026-820 77026-822 77312-950 77312-952
4-Methoxynicotinic acid 95%
4-Methoxynicotinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.1400 g/mol
Boiling Pt: 314.728 °C (760 mm Hg)
Flash Pt: 144.143 °C
Storage Temperature: Ambient
MDL Number: MFCD04115724
CAS Number: 10177-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR