4-Methoxy-1-naphthol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxynaphthalen-1-ol, 1-Hydroxy-4-methoxynaphthalene

A141603-250MG A141603-1G A141603-5G
103803-020EA 19 USD
103803-020 76798-322 77299-226 77299-228
4-Methoxy-1-naphthol 97%
4-Methoxy-1-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
Boiling Pt: 350.1 °C (760 mm Hg)
Melting Pt: 128…130 °C
Flash Pt: 228.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00003976
CAS Number: 84-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR