2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine 96%

Supplier: Ambeed
Warning

A118305-100G A118305-25G
77161-156EA 20 USD
77161-156 77891-845
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine 96%
2-Methoxy-4-methyl-6-(methylamino)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₆H₁₀N₄O
Boiling Pt: 304.9 °C (760 mm Hg)
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00585858
CAS Number: 5248-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR