4-Methoxyfuro[3,2-d]pyrimidine-6-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A101161-100MG
A101161-250MG
103795-832EA
200
USD
103795-832
77093-494
77093-496
4-Methoxyfuro[3,2-d]pyrimidine-6-carboxylic acid 95%
4-Methoxyfuro[3,2-d]pyrimidine-6-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₄ MW: 194.14 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24386867 CAS Number: 1431412-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...