4-Methoxy-3-buten-2-one 90%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybut-3-en-2-one

A467808-25G A467808-5G A467808-100G A467808-10G
103839-066EA 14 USD
103839-066 76842-976 76842-978 77291-842 77291-844
4-Methoxy-3-buten-2-one 90%
4-Methoxy-3-buten-2-one

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.117 g/mol
Boiling Pt: 200 °C (760 mm Hg)
Flash Pt: 63.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008778
CAS Number: 4652-27-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR