4-Methoxybenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: 4-(Aminomethyl)anisole, p-Anisylamine

A944169-25G A944169-10G A944169-5G A944169-100G
103810-608EA 117 USD
103810-608 76845-704 77106-324 77106-326 77686-972
4-Methoxybenzylamine 98%
4-Methoxybenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 236.5 °C (760 mm Hg)
Melting Pt: 10 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008122
CAS Number: 2393-23-9
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR