4-Methoxybenzonitrile 98%

Supplier: Ambeed
Warning

A117453-100G A117453-10G A117453-25G A117453-500G
103824-776EA 8 USD
103824-776 77108-342 77108-344 77108-346 77108-348
4-Methoxybenzonitrile 98%
4-Methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₇NO
Boiling Pt: 256…257 °C (760 mm Hg)
Melting Pt: 57…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00001818
CAS Number: 874-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR