4-Methoxybenzamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzimidamide hydrochloride

A116141-10G A116141-250MG A116141-25G A116141-1G A116141-5G
103798-680EA 23 USD
103798-680 103824-544 77161-002 77161-004 77161-006 77311-362 77311-364
4-Methoxybenzamidine hydrochloride 97%
4-Methoxybenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
Boiling Pt: 255.1 °C (760 mm Hg)
Flash Pt: 108.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00466114
CAS Number: 51721-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR