4-Methoxybenzenesulfonamide 98%

Supplier: Ambeed
Warning

A787211-25G A787211-5G A787211-1G
103841-002EA 23 USD
103841-002 76845-254 76845-256 77308-330
4-Methoxybenzenesulfonamide 98%
4-Methoxybenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₉NO₃S
MW: 187.22 g/mol
Boiling Pt: 352.2 °C (760 mm Hg)
Melting Pt: 111…115 °C
Flash Pt: 166.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00025392
CAS Number: 1129-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR