4-Methoxybenzenesulphonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxybenzene-1-sulphonyl chloride

A173424-10G A173424-5G A173424-25G A173424-100G
103809-076EA 362 USD
103809-076 77138-474 77138-476 77138-478 77301-266 77746-142
4-Methoxybenzenesulphonyl chloride 98%
4-Methoxybenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₇ClO₃S
Boiling Pt: 173 °C (760 mm Hg)
Melting Pt: 39…42 °C
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007446
CAS Number: 98-68-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR