4-Methoxybenzamide 97%

Supplier: Ambeed
Warning

A122556-10G A122556-25G A122556-5G
76688-178EA 23 USD
76688-178 76688-180 77312-368
4-Methoxybenzamide 97%
4-Methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₉NO₂
Boiling Pt: 295.8 °C (760 mm Hg)
Melting Pt: 164…167 °C
Flash Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00007995
CAS Number: 3424-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR