Monomethyl succinate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

A112240-1000G A112240-100G A112240-25G A112240-500G A112240-5G
103823-820EA 9 USD
103823-820 76718-138 76718-140 76718-142 76718-144 77284-222
Monomethyl succinate 97%
Monomethyl succinate

Precaución: for Research Use Only

Formula: C₅H₈O₄
MW: 132.11 g/mol
Melting Pt: 54.2…57.6 °C
Flash Pt: 104.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002788
CAS Number: 3878-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR