4-Methoxy-3-nitrobenzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A191871-25G
A191871-5G
A191871-10G
A191871-250MG
A191871-1G
103816-406EA
6
USD
103816-406
103847-008
77084-872
77084-874
77236-930
77236-932
77302-266
4-Methoxy-3-nitrobenzaldehyde 98%
4-Methoxy-3-nitrobenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₇NO₄ Boiling Pt: 334.8 °C (760 mm Hg) Melting Pt: 97…100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00126498 CAS Number: 31680-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...