4-Methoxy-m-toluidine 97%

Supplier: Ambeed
Warning

A131237-100G A131237-25G A131237-1G A131237-5G A131237-10G A131237-250MG
103801-548EA 6 USD
103801-548 77149-624 77298-466 77314-026 77314-028 77901-305 77901-760
4-Methoxy-m-toluidine 97%
4-Methoxy-m-toluidine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
Melting Pt: 57…60 °C
Flash Pt: 106.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07779234
CAS Number: 136-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR