4-Methoxy-3-methylphenylacetone 97%

Supplier: Ambeed
Warning

A133803-100MG A133803-250MG A133803-1G
103801-916EA 462 USD
103801-916 77285-418 77285-420 77314-608
4-Methoxy-3-methylphenylacetone 97%
4-Methoxy-3-methylphenylacetone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210447
CAS Number: 16882-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR