4-Methoxy-3-methylphenol 96%

Supplier: Ambeed
Warning

A234361-250MG A234361-1G
103810-944EA 115 USD
103810-944 103850-698 76874-314 77089-682
4-Methoxy-3-methylphenol 96%
4-Methoxy-3-methylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.16 g/mol
Boiling Pt: 244.7 °C (760 mm Hg)
Flash Pt: 121.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09836708
CAS Number: 14786-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR