4-Methoxy-m-toluic acid 98%

Supplier: Ambeed
Warning

A408277-5G A408277-10G A408277-1G
103827-178EA 7 USD
103827-178 103838-224 76841-730 77115-792 77307-126
4-Methoxy-m-toluic acid 98%
4-Methoxy-m-toluic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Melting Pt: 177…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00270114
CAS Number: 6880-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR