4-Methoxy-3-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A191083-100G A191083-10G A191083-5G A191083-1G A191083-25G
103816-346EA 9 USD
103816-346 103846-948 76690-888 76690-890 77084-776 77302-244 77302-246
4-Methoxy-3-methylbenzaldehyde 97%
4-Methoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
Flash Pt: 112.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00003375
CAS Number: 32723-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR