4-Methoxy-3-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A158997-10G A158997-1G
103805-208EA 6 USD
103805-208 103843-420 77259-524 77319-282
4-Methoxy-3-(trifluoromethyl)benzoic acid 98%
4-Methoxy-3-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
Boiling Pt: 290.1 °C (760 mm Hg)
Flash Pt: 129.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01091014
CAS Number: 213598-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR