4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile 98%

Supplier: Ambeed
Warning

A504923-25G A504923-5G A504923-100G A504923-10G A504923-1G A504923-500G
103839-418EA 7 USD
103839-418 76843-392 76843-394 77144-054 77144-056 77144-058 77747-037
4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile 98%
4-Methoxy-2-oxo-1,2-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O₂
Boiling Pt: 421.8 °C (760 mm Hg)
Melting Pt: 293 °C
Storage Temperature: Ambient
MDL Number: MFCD00975442
CAS Number: 21642-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR