4-Methoxy-2-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A345443-100G
A345443-10G
A345443-25G
A345443-1G
A345443-5G
103825-926EA
12
USD
103825-926
103836-908
76694-044
76694-046
76694-048
76839-970
76839-972
4-Methoxy-2-methylpyridine 98%
4-Methoxy-2-methylpyridine
Precaución: for Research Use Only
Formula:
C₇H₉NO MW: 123.1500 g/mol Boiling Pt: 184.4 °C (760 mm Hg) Flash Pt: 65.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD06253892 CAS Number: 24103-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...