4-Methoxy-2-methylphenylacetonitrile 98%
Supplier: Ambeed
|
Warning
|
A267724-5G
A267724-10G
103812-876EA
12
USD
103812-876
103852-776
77092-524
77289-386
4-Methoxy-2-methylphenylacetonitrile 98%
4-Methoxy-2-methylphenylacetonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO Boiling Pt: 291.4 °C (760 mm Hg) Flash Pt: 122.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD02178388 CAS Number: 262298-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...