4-Methoxy-2-methylbenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A205559-100G
A205559-10G
A205559-25G
A205559-1G
103817-640EA
20
USD
103817-640
103848-306
77287-794
77287-796
77287-798
77302-960
4-Methoxy-2-methylbenzonitrile 97%
4-Methoxy-2-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₉H₉NO Boiling Pt: 274.7 °C (760 mm Hg) Melting Pt: 104 °C Flash Pt: 115.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425692 CAS Number: 21883-13-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...