4-Methoxy-o-toluidine 98%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole

A148516-100G A148516-500G A148516-25G
103842-052EA 6 USD
103842-052 77028-000 77028-002 77028-004 77317-512
4-Methoxy-o-toluidine 98%
4-Methoxy-o-toluidine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 248
Melting Pt: 13…14 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00007735
CAS Number: 102-50-1
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR