4-methoxy-1-naphthoic acid 95%

Supplier: Ambeed
Warning

A222787-5G A222787-1G A222787-10G A222787-100G A222787-250MG
103819-004EA 6 USD
103819-004 103849-882 77088-492 77303-632 77593-444 77883-573 77883-574
4-methoxy-1-naphthoic acid 95%
4-methoxy-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
Boiling Pt: 388.7 °C (760 mm Hg)
Flash Pt: 155.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02575372
CAS Number: 13041-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR