4-Methoxyindole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxy-1H-indole, Indol-4-yl methyl ether

A139857-10G A139857-1G A139857-25G A139857-5G A139857-100G
103802-784EA 15 USD
103802-784 77272-452 77315-884 77315-886 77315-888 77747-986
4-Methoxyindole 98%
4-Methoxyindole

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 306.9 °C (760 mm Hg)
Melting Pt: 69…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00009737
CAS Number: 4837-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR