4-Isopropylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Isopropylbenzeneboronic acid

A105799-100G A105799-10G A105799-25G A105799-1G A105799-5G
103796-946EA 5 USD
103796-946 103822-690 77245-426 77245-428 77245-430 77309-552 77309-554
4-Isopropylphenylboronic acid 95%
4-Isopropylphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Boiling Pt: 285.9 °C (760 mm Hg)
Melting Pt: 142…144 °C
Flash Pt: 126.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01074614
CAS Number: 16152-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR