4-Isopropylbenzoic acid 98%

Supplier: Ambeed
Warning

A255165-100G A255165-10G A255165-500G A255165-25G A255165-5G
103852-040EA 5 USD
103852-040 76692-610 76692-612 76692-614 77304-646 77304-648
4-Isopropylbenzoic acid 98%
4-Isopropylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00002564
CAS Number: 536-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR