2-Amino-4-iodopyridine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Iodopyridin-2-amine

A150225-250MG A150225-1G A150225-5G
103804-140EA 17 USD
103804-140 103842-274 77286-106 77317-780 77317-782
2-Amino-4-iodopyridine 95%
2-Amino-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
MW: 220.01 g/mol
Boiling Pt: 309.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09038163
CAS Number: 552331-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR