4-Iodo-1,2-dihydropyridin-2-one 96%

Supplier: Ambeed
Warning

Synonyms: 4-Iodopyridin-2(1H)-one, 4-Iodo-1H-pyridin-2-one

A150170-1G A150170-25G A150170-250MG
103799-392EA 24 USD
103799-392 103804-136 103842-270 77299-790 77317-778 77748-043
4-Iodo-1,2-dihydropyridin-2-one 96%
4-Iodo-1,2-dihydropyridin-2-one

Precaución: for Research Use Only

Formula: C₅H₄INO
MW: 221 g/mol
Melting Pt: 186…188 °C
Flash Pt: 148.269 °C
Storage Temperature: Ambient
MDL Number: MFCD09702419
CAS Number: 858839-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR