4-Iodotriphenylamine 98%

Supplier: Ambeed
Warning

A126928-100MG A126928-10G A126928-1G A126928-5G
103800-876EA 8 USD
103800-876 76870-548 76870-550 77298-156 77298-158
4-Iodotriphenylamine 98%
4-Iodotriphenylamine

Precaución: for Research Use Only

Formula: C₁₈H₁₄IN
Melting Pt: 107.1…107.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11046352
CAS Number: 38257-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR